Product Details

SNP ID
rs115319051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71715207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGCCACTCTGTCACAGCACCC[G/T]CCCTGGCCTCTGAAGGGAGCCTCCT
Phenotype
MIM: 605516
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf105 PubMed Links

Gene Details

Gene
C10orf105
Gene Name
chromosome 10 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164375.2 1331 UTR 3 NP_001157847.1
NM_001168390.1 1331 UTR 3 NP_001161862.1
XM_011539808.2 1331 UTR 3 XP_011538110.1
XM_011539809.2 1331 UTR 3 XP_011538111.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 1331 Intron NP_001165401.1
NM_001171931.1 1331 Intron NP_001165402.1
NM_001171932.1 1331 Intron NP_001165403.1
NM_001171933.1 1331 Intron NP_001165404.1
NM_001171934.1 1331 Intron NP_001165405.1
NM_001171935.1 1331 Intron NP_001165406.1
NM_001171936.1 1331 Intron NP_001165407.1
NM_022124.5 1331 Intron NP_071407.4
NM_052836.3 1331 Intron NP_443068.1
XM_006717940.3 1331 Intron XP_006718003.1
XM_006717942.3 1331 Intron XP_006718005.1
XM_011540039.2 1331 Intron XP_011538341.1
XM_011540042.2 1331 Intron XP_011538344.1
XM_011540043.2 1331 Intron XP_011538345.1
XM_011540044.2 1331 Intron XP_011538346.1
XM_011540045.2 1331 Intron XP_011538347.1
XM_011540046.2 1331 Intron XP_011538348.1
XM_011540047.2 1331 Intron XP_011538349.1
XM_011540048.2 1331 Intron XP_011538350.1
XM_011540049.2 1331 Intron XP_011538351.1
XM_011540051.2 1331 Intron XP_011538353.1
XM_011540052.2 1331 Intron XP_011538354.1
XM_017016499.1 1331 Intron XP_016871988.1
XM_017016500.1 1331 Intron XP_016871989.1
XM_017016501.1 1331 Intron XP_016871990.1
XM_017016502.1 1331 Intron XP_016871991.1
XM_017016503.1 1331 Intron XP_016871992.1
XM_017016504.1 1331 Intron XP_016871993.1
XM_017016505.1 1331 Intron XP_016871994.1
XM_017016506.1 1331 Intron XP_016871995.1
XM_017016507.1 1331 Intron XP_016871996.1
XM_017016508.1 1331 Intron XP_016871997.1

View Full Product Details