Product Details

SNP ID
rs116092662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:42585844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTTCCATTCAAGTGGTACCCA[C/T]GGATTACCAAGCTATGTCTGCTTCC
Phenotype
MIM: 194522
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF33B PubMed Links

Gene Details

Gene
ZNF33B
Gene Name
zinc finger protein 33B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305033.1 Intron NP_001291962.1
NM_001305035.1 Intron NP_001291964.1
NM_001305036.1 Intron NP_001291965.1
NM_001305037.1 Intron NP_001291966.1
NM_001305038.1 Intron NP_001291967.1
NM_001305039.1 Intron NP_001291968.1
NM_001305040.1 Intron NP_001291969.1
NM_006955.2 Intron NP_008886.1

View Full Product Details