Product Details

SNP ID
rs116299229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73717325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTTTAAGATGTTTAAGATGAT[A/G]CAGCACAGGGAAAAAATGATCTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMS1P4 PubMed Links

Gene Details

Gene
BMS1P4
Gene Name
BMS1, ribosome biogenesis factor pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details