Product Details

SNP ID
rs116708682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126015301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCCTCTTATAATTGGAGGAAGG[A/G]CTAACATGCCAGATCTAAGGATTTG
Phenotype
MIM: 602714 MIM: 611640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM12 PubMed Links

Gene Details

Gene
ADAM12
Gene Name
ADAM metallopeptidase domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288973.1 5112 UTR 3 NP_001275902.1
NM_001288974.1 5112 Intron NP_001275903.1
NM_001288975.1 5112 Intron NP_001275904.1
NM_003474.5 5112 UTR 3 NP_003465.3
NM_021641.4 5112 Intron NP_067673.2
XM_017016705.1 5112 UTR 3 XP_016872194.1
XM_017016706.1 5112 UTR 3 XP_016872195.1
Gene
FANK1
Gene Name
fibronectin type III and ankyrin repeat domains 1
There are no transcripts associated with this gene.

View Full Product Details