Product Details

SNP ID
rs116806817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35127047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCCGCGGCTACCGCATCACA[A/G]CTGACGTGAGGACTACGTGGGGCCG
Phenotype
MIM: 123812 MIM: 603135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREM PubMed Links

Gene Details

Gene
CREM
Gene Name
cAMP responsive element modulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267562.1 207 UTR 5 NP_001254491.1
NM_001267563.1 207 UTR 5 NP_001254492.1
NM_001267564.1 207 Intron NP_001254493.1
NM_001267565.1 207 Intron NP_001254494.1
NM_001267566.1 207 Intron NP_001254495.1
NM_001267567.1 207 Intron NP_001254496.1
NM_001267568.1 207 Intron NP_001254497.1
NM_001267569.1 207 Intron NP_001254498.1
NM_001267570.1 207 Intron NP_001254499.1
NM_001881.3 207 UTR 5 NP_001872.3
NM_181571.2 207 Intron NP_853549.1
NM_182717.1 207 Intron NP_874386.1
NM_182718.1 207 Intron NP_874387.1
NM_182719.1 207 Intron NP_874388.1
NM_182720.1 207 Intron NP_874389.1
NM_182721.1 207 Intron NP_874390.1
NM_182723.1 207 Intron NP_874392.1
NM_182724.1 207 Intron NP_874393.1
NM_182769.2 207 Intron NP_877570.1
NM_182770.2 207 Intron NP_877571.1
NM_182771.1 207 Intron NP_877572.1
NM_182772.1 207 Intron NP_877573.1
NM_183011.1 207 Intron NP_898829.1
NM_183012.1 207 Intron NP_898830.1
NM_183013.2 207 UTR 5 NP_898831.1
NM_183060.2 207 Intron NP_898883.1
XM_006717378.3 207 Intron XP_006717441.1
XM_006717379.3 207 Intron XP_006717442.1
XM_006717382.3 207 Intron XP_006717445.1
XM_006717383.3 207 Intron XP_006717446.1
XM_006717387.3 207 Intron XP_006717450.1
XM_011519316.2 207 Intron XP_011517618.1
XM_011519324.2 207 Intron XP_011517626.1
XM_011519325.2 207 Intron XP_011517627.1
XM_011519327.2 207 Intron XP_011517629.1
XM_011519330.2 207 Intron XP_011517632.1
XM_011519331.2 207 Intron XP_011517633.1
XM_011519332.2 207 Intron XP_011517634.1
XM_011519333.1 207 Intron XP_011517635.1
XM_011519335.2 207 Intron XP_011517637.1
XM_017015716.1 207 UTR 5 XP_016871205.1
XM_017015717.1 207 UTR 5 XP_016871206.1
XM_017015718.1 207 Intron XP_016871207.1
XM_017015719.1 207 UTR 5 XP_016871208.1
XM_017015720.1 207 Intron XP_016871209.1
XM_017015721.1 207 UTR 5 XP_016871210.1
XM_017015722.1 207 Intron XP_016871211.1
XM_017015723.1 207 UTR 5 XP_016871212.1
XM_017015724.1 207 UTR 5 XP_016871213.1
XM_017015725.1 207 Intron XP_016871214.1
XM_017015726.1 207 Intron XP_016871215.1
XM_017015727.1 207 UTR 5 XP_016871216.1
XM_017015728.1 207 UTR 5 XP_016871217.1
XM_017015729.1 207 UTR 5 XP_016871218.1
XM_017015730.1 207 Intron XP_016871219.1
XM_017015731.1 207 Intron XP_016871220.1
XM_017015732.1 207 Intron XP_016871221.1
XM_017015733.1 207 Intron XP_016871222.1
XM_017015734.1 207 UTR 5 XP_016871223.1
XM_017015735.1 207 Intron XP_016871224.1
XM_017015736.1 207 Intron XP_016871225.1
XM_017015737.1 207 UTR 5 XP_016871226.1
Gene
CUL2
Gene Name
cullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198777.1 207 Intron NP_001185706.1
NM_001198778.1 207 Intron NP_001185707.1
NM_001198779.1 207 Intron NP_001185708.1
NM_001324375.1 207 Intron NP_001311304.1
NM_001324376.1 207 Intron NP_001311305.1
NM_003591.3 207 Intron NP_003582.2
XM_011519743.1 207 Intron XP_011518045.1
XM_011519744.1 207 Intron XP_011518046.1
XM_011519745.1 207 Intron XP_011518047.1
XM_011519747.1 207 Intron XP_011518049.1

View Full Product Details