Product Details

SNP ID
rs138012316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122698026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTACATTCATTTTTATTTCTC[G/T]TCTTTTTGTGTTTTTTCCCTCTGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 747 Silent Mutation NP_001010912.1

View Full Product Details