Product Details
- SNP ID
-
rs138249221
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:45618184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTCCTGTTTTCTTTCCACAAAG[A/C]AAACAATGATTTGTTGTTTTCTTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZFAND4
PubMed Links
Gene Details
- Gene
- ZFAND4
- Gene Name
- zinc finger AN1-type containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128324.2 |
2706 |
Missense Mutation |
TTG,TTT |
L668F |
NP_001121796.1 |
NM_001282905.1 |
2706 |
Missense Mutation |
TTG,TTT |
L594F |
NP_001269834.1 |
NM_001282906.1 |
2706 |
Missense Mutation |
TTG,TTT |
L594F |
NP_001269835.1 |
NM_174890.3 |
2706 |
Missense Mutation |
TTG,TTT |
L668F |
NP_777550.2 |
XM_005271842.2 |
2706 |
Missense Mutation |
TTG,TTT |
L569F |
XP_005271899.1 |
XM_005271843.2 |
2706 |
Missense Mutation |
TTG,TTT |
L569F |
XP_005271900.1 |
XM_005271844.3 |
2706 |
Missense Mutation |
TTG,TTT |
L550F |
XP_005271901.1 |
XM_011540366.2 |
2706 |
Missense Mutation |
TTG,TTT |
L676F |
XP_011538668.1 |
XM_011540368.1 |
2706 |
Missense Mutation |
TTG,TTT |
L496F |
XP_011538670.1 |
XM_017016931.1 |
2706 |
Missense Mutation |
TTG,TTT |
L682F |
XP_016872420.1 |
XM_017016932.1 |
2706 |
Missense Mutation |
TTG,TTT |
L674F |
XP_016872421.1 |
XM_017016933.1 |
2706 |
Missense Mutation |
TTG,TTT |
L674F |
XP_016872422.1 |
XM_017016934.1 |
2706 |
Missense Mutation |
TTG,TTT |
L674F |
XP_016872423.1 |
XM_017016935.1 |
2706 |
UTR 3 |
|
|
XP_016872424.1 |
XM_017016936.1 |
2706 |
Missense Mutation |
TTG,TTT |
L626F |
XP_016872425.1 |
XM_017016937.1 |
2706 |
Missense Mutation |
TTG,TTT |
L626F |
XP_016872426.1 |
XM_017016938.1 |
2706 |
Missense Mutation |
TTG,TTT |
L620F |
XP_016872427.1 |
XM_017016939.1 |
2706 |
Missense Mutation |
TTG,TTT |
L620F |
XP_016872428.1 |
XM_017016940.1 |
2706 |
Missense Mutation |
TTG,TTT |
L600F |
XP_016872429.1 |
XM_017016941.1 |
2706 |
Missense Mutation |
TTG,TTT |
L600F |
XP_016872430.1 |
XM_017016942.1 |
2706 |
Missense Mutation |
TTG,TTT |
L575F |
XP_016872431.1 |
XM_017016943.1 |
2706 |
Missense Mutation |
TTG,TTT |
L575F |
XP_016872432.1 |
XM_017016944.1 |
2706 |
Missense Mutation |
TTG,TTT |
L575F |
XP_016872433.1 |
XM_017016945.1 |
2706 |
Missense Mutation |
TTG,TTT |
L556F |
XP_016872434.1 |
XM_017016946.1 |
2706 |
Missense Mutation |
TTG,TTT |
L475F |
XP_016872435.1 |
View Full Product Details