Product Details
- SNP ID
-
rs138261581
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47348841 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAACTGCTTGCCTGGCTGCAAAG[G/A]GGCCTCCGCCATGAGGTTCTGGAGG
- Phenotype
-
MIM: 180290
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBP3
PubMed Links
Gene Details
- Gene
- RBP3
- Gene Name
- retinol binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002900.2 |
471 |
Silent Mutation |
AGA,AGG |
R119R |
NP_002891.1 |
View Full Product Details