Product Details
- SNP ID
-
rs138300341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127878412 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAACAGTATCATGGTCACAAAAA[A/C]GAAGGATGCTGAAAGGAAGATGGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLRN3
PubMed Links
Gene Details
- Gene
- CLRN3
- Gene Name
- clarin 3
View Full Product Details