Product Details
- SNP ID
-
hCV160322931
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:116591930 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAACCCAAACAACTTTCAAGTGA[C/G]ACCTCTGTCATTTAAATGTCACTGT
- Phenotype
-
MIM: 604422
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PNLIPRP1
PubMed Links
Gene Details
- Gene
- PNLIPRP1
- Gene Name
- pancreatic lipase related protein 1
There are no transcripts associated with this gene.
View Full Product Details