Product Details

SNP ID
rs138894690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1019574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGGTCACGTCATCCAGGTGAGG[A/C]CACCACCGGTACAGAAACCTCTCGG
Phenotype
MIM: 615389 MIM: 615391
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP4 PubMed Links

Gene Details

Gene
GTPBP4
Gene Name
GTP binding protein 4
There are no transcripts associated with this gene.

Gene
IDI2
Gene Name
isopentenyl-diphosphate delta isomerase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033261.2 692 Missense Mutation TGG,TGT W209C NP_150286.1
Gene
IDI2-AS1
Gene Name
IDI2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details