Product Details

SNP ID
rs140088745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122461989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCAGGCCGGCCTCTGTGTGTGC[G/T]CCAGCAGCGAGCCGGTGTGCGGCAG
Phenotype
MIM: 611313 MIM: 602194
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMS2 PubMed Links

Gene Details

Gene
ARMS2
Gene Name
age-related maculopathy susceptibility 2
There are no transcripts associated with this gene.

Gene
HTRA1
Gene Name
HtrA serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002775.4 465 Missense Mutation GCC,TCC A113S NP_002766.1

View Full Product Details