Product Details
- SNP ID
-
rs140396238
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:101579728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGATGATGTCCAGGCGCCGGTGCC[A/G]CCGCCCTGGCCCTGGGAGCCGGCAC
- Phenotype
-
MIM: 616467
MIM: 606343
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DPCD
PubMed Links
Gene Details
- Gene
- DPCD
- Gene Name
- deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.
- Gene
- POLL
- Gene Name
- polymerase (DNA) lambda
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001174084.1 |
1948 |
Missense Mutation |
CGG,TGG |
R485W |
NP_001167555.1 |
NM_001174085.1 |
1948 |
Missense Mutation |
CGG,TGG |
R393W |
NP_001167556.1 |
NM_001308382.1 |
1948 |
Missense Mutation |
CGG,TGG |
R210W |
NP_001295311.1 |
NM_013274.3 |
1948 |
Missense Mutation |
CGG,TGG |
R485W |
NP_037406.1 |
XM_006717775.1 |
1948 |
Missense Mutation |
CGG,TGG |
R224W |
XP_006717838.1 |
XM_006717776.1 |
1948 |
Missense Mutation |
CGG,TGG |
R224W |
XP_006717839.1 |
XM_006717777.1 |
1948 |
Missense Mutation |
CGG,TGG |
R210W |
XP_006717840.1 |
XM_011539650.1 |
1948 |
Missense Mutation |
CGG,TGG |
R492W |
XP_011537952.1 |
XM_011539651.1 |
1948 |
Missense Mutation |
CGG,TGG |
R485W |
XP_011537953.1 |
XM_011539652.1 |
1948 |
Missense Mutation |
CGG,TGG |
R492W |
XP_011537954.1 |
XM_011539653.1 |
1948 |
Missense Mutation |
CGG,TGG |
R492W |
XP_011537955.1 |
XM_011539654.1 |
1948 |
Missense Mutation |
CGG,TGG |
R400W |
XP_011537956.1 |
XM_011539655.1 |
1948 |
Missense Mutation |
CGG,TGG |
R393W |
XP_011537957.1 |
XM_011539656.1 |
1948 |
Missense Mutation |
CGG,TGG |
R386W |
XP_011537958.1 |
XM_011539657.1 |
1948 |
Missense Mutation |
CGG,TGG |
R404W |
XP_011537959.1 |
XM_011539659.1 |
1948 |
Missense Mutation |
CGG,TGG |
R323W |
XP_011537961.1 |
XM_011539660.1 |
1948 |
Missense Mutation |
CGG,TGG |
R323W |
XP_011537962.1 |
XM_011539662.1 |
1948 |
Missense Mutation |
CGG,TGG |
R231W |
XP_011537964.1 |
XM_011539663.1 |
1948 |
Missense Mutation |
CGG,TGG |
R231W |
XP_011537965.1 |
XM_011539664.1 |
1948 |
Missense Mutation |
CGG,TGG |
R217W |
XP_011537966.1 |
XM_011539665.2 |
1948 |
Missense Mutation |
CGG,TGG |
R217W |
XP_011537967.1 |
XM_011539666.1 |
1948 |
Missense Mutation |
CGG,TGG |
R217W |
XP_011537968.1 |
XM_011539667.1 |
1948 |
Missense Mutation |
CGG,TGG |
R177W |
XP_011537969.1 |
XM_017016084.1 |
1948 |
Missense Mutation |
CGG,TGG |
R391W |
XP_016871573.1 |
XM_017016085.1 |
1948 |
Missense Mutation |
CGG,TGG |
R384W |
XP_016871574.1 |
XM_017016086.1 |
1948 |
Missense Mutation |
CGG,TGG |
R379W |
XP_016871575.1 |
XM_017016087.1 |
1948 |
Missense Mutation |
CGG,TGG |
R299W |
XP_016871576.1 |
XM_017016088.1 |
1948 |
UTR 3 |
|
|
XP_016871577.1 |
XM_017016089.1 |
1948 |
Missense Mutation |
CGG,TGG |
R323W |
XP_016871578.1 |
XM_017016090.1 |
1948 |
UTR 3 |
|
|
XP_016871579.1 |
XM_017016091.1 |
1948 |
Missense Mutation |
CGG,TGG |
R316W |
XP_016871580.1 |
XM_017016092.1 |
1948 |
UTR 3 |
|
|
XP_016871581.1 |
XM_017016093.1 |
1948 |
Missense Mutation |
CGG,TGG |
R217W |
XP_016871582.1 |
View Full Product Details