Product Details
- SNP ID
-
rs140729995
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92461246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGTCTTTAAATATGCAACCTCA[A/G]TGTTATCTGAAAAAGTAATCAAACA
- Phenotype
-
MIM: 146680
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IDE
PubMed Links
Gene Details
- Gene
- IDE
- Gene Name
- insulin degrading enzyme
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001165946.1 |
1428 |
Missense Mutation |
ACT,ATT |
T368I |
NP_001159418.1 |
NM_001322793.1 |
1428 |
Missense Mutation |
ACT,ATT |
T923I |
NP_001309722.1 |
NM_001322794.1 |
1428 |
Missense Mutation |
ACT,ATT |
T884I |
NP_001309723.1 |
NM_001322795.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
NP_001309724.1 |
NM_001322796.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
NP_001309725.1 |
NM_001322797.1 |
1428 |
Missense Mutation |
ACT,ATT |
T368I |
NP_001309726.1 |
NM_004969.3 |
1428 |
Missense Mutation |
ACT,ATT |
T923I |
NP_004960.2 |
XM_017016187.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
XP_016871676.1 |
XM_017016188.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
XP_016871677.1 |
XM_017016189.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
XP_016871678.1 |
XM_017016190.1 |
1428 |
Missense Mutation |
ACT,ATT |
T882I |
XP_016871679.1 |
View Full Product Details