Product Details
- SNP ID
-
rs140930248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:113866232 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAAAAAGCAAAACCTACTTAATA[A/G]GAATGAATTGTTCTGATGTTTAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NHLRC2
PubMed Links
Gene Details
- Gene
- NHLRC2
- Gene Name
- NHL repeat containing 2
View Full Product Details