Product Details

SNP ID
rs141806369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46009425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTACCCACCTGTAGAGGAGTTC[G/A]ATATAACCACTTCTCTTTATCAACT
Phenotype
MIM: 601984 MIM: 605034
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NCOA4 PubMed Links

Gene Details

Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145260.1 Intron NP_001138732.1
NM_001145261.1 Intron NP_001138733.1
NM_001145262.1 Intron NP_001138734.1
NM_001145263.1 Intron NP_001138735.1
NM_005437.3 Intron NP_005428.1
Gene
TIMM23
Gene Name
translocase of inner mitochondrial membrane 23
There are no transcripts associated with this gene.

View Full Product Details