Product Details

SNP ID
rs141891824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91629397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGGTTATTTAGCTTTTCTCCA[C/T]AAGGATACGTAGAATGGATCTTTTT
Phenotype
MIM: 602999
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R3C PubMed Links

Gene Details

Gene
PPP1R3C
Gene Name
protein phosphatase 1 regulatory subunit 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005398.5 1616 UTR 3 NP_005389.1

View Full Product Details