Product Details

SNP ID
rs142789275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73782186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTAACCGCGTCGGGGAGCGGCG[C/T]CGGGAGAAGGGCGGTGAGCAGTCGG
Phenotype
MIM: 608842 MIM: 616932
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD1 PubMed Links

Gene Details

Gene
CHCHD1
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203298.2 137 Silent Mutation CGC,CGT R37R NP_976043.1
Gene
FUT11
Gene Name
fucosyltransferase 11
There are no transcripts associated with this gene.

Gene
ZSWIM8
Gene Name
zinc finger SWIM-type containing 8
There are no transcripts associated with this gene.

View Full Product Details