Product Details

SNP ID
rs143123794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122698069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGCCTCCTCTTTCTTATCTTG[A/G]TTGGCATCATCACAATTATGGGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 704 Silent Mutation AAC,AAT N224N NP_001010912.1

View Full Product Details