Product Details

SNP ID
rs143137940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102231762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAACCGTCCTCTGGGGAGCCGCC[A/G]GGCAGCGAAGCCGAGGCCTTTTCTG
Phenotype
MIM: 611815 MIM: 602669
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELOVL3 PubMed Links

Gene Details

Gene
ELOVL3
Gene Name
ELOVL fatty acid elongase 3
There are no transcripts associated with this gene.

Gene
PITX3
Gene Name
paired like homeodomain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005029.3 301 Silent Mutation NP_005020.1

View Full Product Details