Product Details

SNP ID
rs143159665
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:65915927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTTTGAGATGACTAAGATAGC[A/G]TTAGTAGCCTCATTCAAAACATTTT
Phenotype
MIM: 607667
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CTNNA3 PubMed Links

Gene Details

Gene
CTNNA3
Gene Name
catenin alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127384.2 7288 UTR 3 NP_001120856.1
NM_001291133.1 7288 Intron NP_001278062.1
NM_013266.3 7288 UTR 3 NP_037398.2
XM_017016151.1 7288 UTR 3 XP_016871640.1
XM_017016152.1 7288 UTR 3 XP_016871641.1
XM_017016153.1 7288 UTR 3 XP_016871642.1
XM_017016154.1 7288 UTR 3 XP_016871643.1
XM_017016155.1 7288 UTR 3 XP_016871644.1
XM_017016156.1 7288 UTR 3 XP_016871645.1
XM_017016157.1 7288 UTR 3 XP_016871646.1
XM_017016158.1 7288 Intron XP_016871647.1

View Full Product Details