Product Details

SNP ID
rs143355899
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:63535709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTTGCAAAGAATCTGATATGA[C/T]AGTGGTAGAATTCAAAATCAATGGG
Phenotype
MIM: 609348
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
REEP3 PubMed Links

Gene Details

Gene
REEP3
Gene Name
receptor accessory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001330.2 Intron NP_001001330.1
XM_011539501.2 Intron XP_011537803.1
XM_017015896.1 Intron XP_016871385.1

View Full Product Details