Product Details

SNP ID
rs143383772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46003252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCTGACAGGACTAACACTTAC[T/C]AGCCTCTATGCACTATATAATAACT
Phenotype
MIM: 601984 MIM: 605034
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
NCOA4 PubMed Links

Gene Details

Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

Gene
TIMM23
Gene Name
translocase of inner mitochondrial membrane 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006327.3 736 Silent Mutation NP_006318.1
XM_011539098.1 736 Intron XP_011537400.1
XM_011539099.1 736 Intron XP_011537401.1

View Full Product Details