Product Details

SNP ID
rs144120149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69453177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGGATGGAGCAAGGACCTGTTG[G/T]TGGAAGCTCTAGGGAGACAGAAGGT
Phenotype
MIM: 613140
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TSPAN15 PubMed Links

Gene Details

Gene
TSPAN15
Gene Name
tetraspanin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012339.3 Intron NP_036471.1
XM_005269666.4 Intron XP_005269723.1
XM_005269667.3 Intron XP_005269724.1
XM_011539562.2 Intron XP_011537864.1
XM_011539563.2 Intron XP_011537865.1
XM_017016010.1 Intron XP_016871499.1

View Full Product Details