Product Details

SNP ID
rs144739725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103455510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCAGGGGCCGTGGCCAGTGTCC[C/T]GTGGGTGTGACCCAGCTCCAGGTCA
Phenotype
MIM: 612234 MIM: 612235 MIM: 612333
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALHM1 PubMed Links

Gene Details

Gene
CALHM1
Gene Name
calcium homeostasis modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001412.3 933 Missense Mutation AGG,GGG R265G NP_001001412.3
Gene
CALHM2
Gene Name
calcium homeostasis modulator 2
There are no transcripts associated with this gene.

Gene
PDCD11
Gene Name
programmed cell death 11
There are no transcripts associated with this gene.

View Full Product Details