Product Details

SNP ID
rs144870790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101227340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGCTGCTGGCCGGCTGGTCC[A/G]TGAGCGGAGAGGCAGGCGAGAGCTG
Phenotype
MIM: 604255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LBX1 PubMed Links

Gene Details

Gene
LBX1
Gene Name
ladybird homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006562.4 921 Missense Mutation ACG,ATG T259M NP_006553.2
Gene
LBX1-AS1
Gene Name
LBX1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details