Product Details

SNP ID
rs145685617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132445697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGATTGCGGCGTTTCTCTACAGC[G/T]TGTCCATGGGCCCAGTGCCTGGCGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf91 PubMed Links

Gene Details

Gene
C10orf91
Gene Name
chromosome 10 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173541.2 71 Missense Mutation GTG,TTG V11L NP_775812.1
Gene
LOC105378568
Gene Name
uncharacterized LOC105378568
There are no transcripts associated with this gene.

View Full Product Details