Product Details

SNP ID
rs145887258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99533371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGAACTCACTAGCCGCAGCAGA[C/T]GGCCACGGGGATTCAGGGCTGTGTC
Phenotype
MIM: 606727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01475 PubMed Links

Gene Details

Gene
LINC01475
Gene Name
long intergenic non-protein coding RNA 1475
There are no transcripts associated with this gene.

Gene
NKX2-3
Gene Name
NK2 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145285.2 439 Silent Mutation GAC,GAT D80D NP_660328.2
XM_011539370.1 439 Silent Mutation GAC,GAT D80D XP_011537672.1

View Full Product Details