Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_080722.4 | 563 | Missense Mutation | GAA,GGA | E93G | NP_542453.2 |
NM_139155.3 | 563 | Missense Mutation | GAA,GGA | E93G | NP_631894.2 |
XM_011539300.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537602.1 |
XM_011539301.2 | 563 | Intron | XP_011537603.1 | ||
XM_011539302.2 | 563 | Intron | XP_011537604.1 | ||
XM_011539303.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537605.1 |
XM_011539305.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537607.1 |
XM_011539306.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537608.1 |
XM_011539307.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537609.1 |
XM_011539308.2 | 563 | Missense Mutation | GAA,GGA | E93G | XP_011537610.1 |
XM_011539309.1 | 563 | Intron | XP_011537611.1 |