Product Details
- SNP ID
-
rs146172810
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29289372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATTCTAAGGCAGGGGATGGAAA[C/T]CAACTCTATTTGTAAGCTCTGTATC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL1
PubMed Links
Gene Details
- Gene
- LYZL1
- Gene Name
- lysozyme like 1
View Full Product Details