Product Details

SNP ID
hCV160692165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46003236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGATAGCACGAGGTGGTCTGA[T/C]AGGACTAACACTTACCAGCCTCTAT
Phenotype
MIM: 601984 MIM: 605034
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
NCOA4 PubMed Links

Gene Details

Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

Gene
TIMM23
Gene Name
translocase of inner mitochondrial membrane 23
There are no transcripts associated with this gene.

View Full Product Details