Product Details

SNP ID
rs146761373
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102231997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCCCTTGCAGCCGTGCTCTGG[C/G]AGCTGGGGGTGCGGAGTGCCAGCGT
Phenotype
MIM: 611815 MIM: 602669
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ELOVL3 PubMed Links

Gene Details

Gene
ELOVL3
Gene Name
ELOVL fatty acid elongase 3
There are no transcripts associated with this gene.

Gene
PITX3
Gene Name
paired like homeodomain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005029.3 238 Silent Mutation CTC,CTG L28L NP_005020.1

View Full Product Details