Product Details

SNP ID
rs147054734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122698095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCATCATCACAATTATGGGTGT[C/T]ATAGTTGTCTTCCTTTCGTCTGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 678 Missense Mutation AAC,GAC N216D NP_001010912.1

View Full Product Details