Product Details

SNP ID
rs147473024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72059791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGCTCCCAGGCCATGCTTCC[G/T]TCTCCATCACCCTTGGACCCTCTCT
Phenotype
MIM: 607988
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPOCK2 PubMed Links

Gene Details

Gene
SPOCK2
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134434.1 4689 Intron NP_001127906.1
NM_001244950.1 4689 UTR 3 NP_001231879.1
NM_014767.2 4689 UTR 3 NP_055582.1
XM_011540404.2 4689 UTR 3 XP_011538706.1
XM_017016985.1 4689 Intron XP_016872474.1

View Full Product Details