Product Details

SNP ID
rs147501142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125839186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCACGACACCACTTTTCCACAC[A/G]GTCTAGGAGGGAAAGAACACAAGGC
Phenotype
MIM: 611883 MIM: 607960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCCIP PubMed Links

Gene Details

Gene
BCCIP
Gene Name
BRCA2 and CDKN1A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016567.3 2209 Intron NP_057651.1
NM_078468.2 2209 Intron NP_510868.1
NM_078469.2 2209 Intron NP_510869.1
Gene
DHX32
Gene Name
DEAH-box helicase 32 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018180.2 2209 Missense Mutation CGT,TGT R566C NP_060650.2
XM_017016404.1 2209 Missense Mutation CGT,TGT R566C XP_016871893.1
XM_017016405.1 2209 Missense Mutation CGT,TGT R566C XP_016871894.1

View Full Product Details