Product Details

SNP ID
rs147696485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127878299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTATGAGCCAGAACGAGTATCC[A/G]TAACTGTGGGTCGTTCCTTTACTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLRN3 PubMed Links

Gene Details

Gene
CLRN3
Gene Name
clarin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152311.3 513 Silent Mutation TAC,TAT Y177Y NP_689524.1
XM_011539274.2 513 Silent Mutation TAC,TAT Y117Y XP_011537576.1

View Full Product Details