Product Details

SNP ID
rs148744638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70140009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCAGCAGCGCGGTCCAGCTCA[C/T]GGAGGCCACCTAGGTCTTGGGTCTG
Phenotype
MIM: 605159 MIM: 611017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIFM2 PubMed Links

Gene Details

Gene
AIFM2
Gene Name
apoptosis inducing factor, mitochondria associated 2
There are no transcripts associated with this gene.

Gene
TYSND1
Gene Name
trypsin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040273.2 1770 UTR 3 NP_001035363.1
NM_173555.3 1770 Missense Mutation CAT,CGT H539R NP_775826.2

View Full Product Details