Product Details

SNP ID
rs149042847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69572863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGTCCCCCGCGCCGTGCCCGGA[A/G]CTTCCTCGGGGCCCCTCGGCAGCCT
Phenotype
MIM: 604882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEUROG3 PubMed Links

Gene Details

Gene
NEUROG3
Gene Name
neurogenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020999.3 464 Missense Mutation CTC,TTC L61F NP_066279.2
XM_017016280.1 464 Missense Mutation CTC,TTC L61F XP_016871769.1

View Full Product Details