Product Details

SNP ID
rs149072229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94775130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCATGGTGGTGCTGCATGGATAT[A/G]AAGTGGTGAAGGAAGCCCTGATTGA
Phenotype
MIM: 124020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2C19 PubMed Links

Gene Details

Gene
CYP2C19
Gene Name
cytochrome P450 family 2 subfamily C member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000769.2 266 Missense Mutation AAA,GAA K81E NP_000760.1

View Full Product Details