Product Details

SNP ID
rs149124655
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116664171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAAGGAAAGAGACATTTCTCCT[C/G]ACCTGGAATAGAAAAATCGGGATAT
Phenotype
MIM: 610701
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
C10orf82 PubMed Links

Gene Details

Gene
C10orf82
Gene Name
chromosome 10 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011539340.2 Intron XP_011537642.1
XM_011539341.1 Intron XP_011537643.1
XM_011539342.2 Intron XP_011537644.1
XM_011539343.2 Intron XP_011537645.1
XM_011539344.2 Intron XP_011537646.1
XM_011539345.2 Intron XP_011537647.1
XM_011539346.2 Intron XP_011537648.1
XM_017015753.1 Intron XP_016871242.1
XM_017015754.1 Intron XP_016871243.1
XM_017015755.1 Intron XP_016871244.1
Gene
HSPA12A
Gene Name
heat shock protein family A (Hsp70) member 12A
There are no transcripts associated with this gene.

Gene
LOC105378499
Gene Name
uncharacterized LOC105378499
There are no transcripts associated with this gene.

View Full Product Details