Product Details

SNP ID
rs149226838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56358939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAACCTCCGCCAGATGCTGCAG[A/C]TGCTTCTCCTGCCAGGAGTGAGCAT
Phenotype
MIM: 609177
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 526 Missense Mutation CAG,CAT Q163H NP_001005413.1
NM_007057.3 526 Missense Mutation CAG,CAT Q163H NP_008988.2
NM_032997.2 526 Missense Mutation CAG,CAT Q163H NP_127490.1
XM_017015605.1 526 Missense Mutation CAG,CAT Q163H XP_016871094.1

View Full Product Details