Product Details

SNP ID
rs149242882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35640499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCGAGAGGAAGATAATGGGCC[G/T]CTCCGGGTACTTGAAGCGCTCCATG
Phenotype
MIM: 606146
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FZD8 PubMed Links

Gene Details

Gene
FZD8
Gene Name
frizzled class receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031866.2 936 Silent Mutation AGG,CGG R311R NP_114072.1
Gene
MIR4683
Gene Name
microRNA 4683
There are no transcripts associated with this gene.

View Full Product Details