Product Details

SNP ID
rs149293567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73632121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGGGCATCTGGAAGGTCATGC[A/G]TTTGGAGGCCTTCTCATATCCACCA
Phenotype
MIM: 605603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYOZ1 PubMed Links

Gene Details

Gene
MYOZ1
Gene Name
myozenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021245.3 1074 Missense Mutation CGC,TGC R237C NP_067068.1
Gene
USP54
Gene Name
ubiquitin specific peptidase 54
There are no transcripts associated with this gene.

View Full Product Details