Product Details
- SNP ID
-
rs149719025
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:132333730 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGAGGTCTTTAGAATCCCCAGCC[G/T]TCAAGTAAGTGGGGCTGCTCCTCAG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC27
PubMed Links
Gene Details
- Gene
- LRRC27
- Gene Name
- leucine rich repeat containing 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143757.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
NP_001137229.1 |
NM_001143758.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
NP_001137230.1 |
NM_001143759.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
NP_001137231.1 |
NM_001309474.1 |
347 |
Intron |
|
|
NP_001296403.1 |
NM_030626.2 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
NP_085129.1 |
XM_006717983.2 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_006718046.1 |
XM_006717984.2 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_006718047.1 |
XM_011540208.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_011538510.1 |
XM_011540209.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_011538511.1 |
XM_011540210.2 |
347 |
Intron |
|
|
XP_011538512.1 |
XM_017016690.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_016872179.1 |
XM_017016691.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_016872180.1 |
XM_017016692.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_016872181.1 |
XM_017016693.1 |
347 |
Missense Mutation |
CGT,CTT |
R69L |
XP_016872182.1 |
XM_017016694.1 |
347 |
UTR 5 |
|
|
XP_016872183.1 |
- Gene
- STK32C
- Gene Name
- serine/threonine kinase 32C
There are no transcripts associated with this gene.
View Full Product Details