Product Details

SNP ID
rs149782162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5196915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCTAAATGATGGTCACTTCAT[A/G]CCCGTATTGGGATTTGGCACCTATG
Phenotype
MIM: 600451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR1C4 PubMed Links

Gene Details

Gene
AKR1C4
Gene Name
aldo-keto reductase family 1, member C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001818.3 81 Missense Mutation ATA,ATG I16M NP_001809.3

View Full Product Details