Product Details

SNP ID
rs150807488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96982150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGTGCAGCTTCCCAGAGAAGAC[A/C]ACCCTGAAGAACCTAGCAAGGAAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 867 Missense Mutation AAC,CAC N254H NP_056467.2

View Full Product Details