Product Details

SNP ID
rs150815977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101131595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGGTCACGCAGAGCCCATTAG[C/T]TTCGGCATCGACCAGATCCTCAACA
Phenotype
MIM: 186770 MIM: 612734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLX1 PubMed Links

Gene Details

Gene
TLX1
Gene Name
T-cell leukemia homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195517.1 292 Silent Mutation AGC,AGT S18S NP_001182446.1
NM_005521.3 292 Silent Mutation AGC,AGT S18S NP_005512.1
XM_011539744.2 292 Silent Mutation AGC,AGT S18S XP_011538046.1
XM_011539745.2 292 Silent Mutation AGC,AGT S18S XP_011538047.1
Gene
TLX1NB
Gene Name
TLX1 neighbor
There are no transcripts associated with this gene.

View Full Product Details