Product Details

SNP ID
rs150846674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27398355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCTGTTGTCTGAATGAAGCCC[C/T]GGGAAGAAATGATTTCATTTGATGA
Phenotype
MIM: 611791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTCHD3 PubMed Links

Gene Details

Gene
PTCHD3
Gene Name
patched domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034842.3 2361 Missense Mutation CAG,CGG Q748R NP_001030014.2

View Full Product Details