Product Details
- SNP ID
-
rs150947060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92461238 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTTGGTAAGTGTCTTTAAATATG[C/T]AACCTCAGTGTTATCTGAAAAAGTA
- Phenotype
-
MIM: 146680
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IDE
PubMed Links
Gene Details
- Gene
- IDE
- Gene Name
- insulin degrading enzyme
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001165946.1 |
1436 |
Missense Mutation |
ACA,GCA |
T371A |
NP_001159418.1 |
NM_001322793.1 |
1436 |
Missense Mutation |
ACA,GCA |
T926A |
NP_001309722.1 |
NM_001322794.1 |
1436 |
Missense Mutation |
ACA,GCA |
T887A |
NP_001309723.1 |
NM_001322795.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
NP_001309724.1 |
NM_001322796.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
NP_001309725.1 |
NM_001322797.1 |
1436 |
Missense Mutation |
ACA,GCA |
T371A |
NP_001309726.1 |
NM_004969.3 |
1436 |
Missense Mutation |
ACA,GCA |
T926A |
NP_004960.2 |
XM_017016187.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
XP_016871676.1 |
XM_017016188.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
XP_016871677.1 |
XM_017016189.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
XP_016871678.1 |
XM_017016190.1 |
1436 |
Missense Mutation |
ACA,GCA |
T885A |
XP_016871679.1 |
View Full Product Details