Product Details

SNP ID
rs112710258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62715380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTGGTAGTAGTAACTCTGCCA[A/C]TAGAAGAGAGGGAAACCCCATCACT
Phenotype
MIM: 606158 MIM: 608941
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BSCL2 PubMed Links

Gene Details

Gene
BSCL2
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
There are no transcripts associated with this gene.

Gene
GNG3
Gene Name
G protein subunit gamma 3
There are no transcripts associated with this gene.

Gene
HNRNPUL2
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079559.2 Intron NP_001073027.1
Gene
HNRNPUL2-BSCL2
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details